Learn More
Enzo Life Sciences Sparfloxacin (5g). CAS: 110871-86-8
SDP
Supplier:  Enzo Life Sciences ALX380302G005
Encompass_Preferred
Fluoroquinolone with broad spectrum antibacterial activity against Gram-positive and Gram-negative bacteria. Inhibits bacterial DNA gyrase (topoisomerase). Effective against Mycobacterium tuberculosis. Shows photosensitizing effects. Alternative name: rel-5-Amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethyl-1-piperazinyl]-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, AT-4140. Purity: ≥98% (Assay). Solubility: Soluble in 100% ethanol or water. Long Term Storage: -20°C.
Catalog No. 50-200-9025