Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sparfloxacin, G-Biosciences
SCI_ED

Catalog No. S000022
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000022 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000022 Supplier G-Biosciences Supplier No. RC1100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sparfloxacin is a broad-spectrum fluoroquinolone antibiotic targeting gram-negative and gram-positive bacteria as well as Mycoplasma species

It is especially effective against bacteria responsible for respiratory tract infections such as pneumonia. Sparfloxacin also displays photosensitizing effects.

Chemical Identifiers

Molecular Weight (g/mol) 392.4

Specifications

Physical Form Powder
Quantity 10 g
Product Line Gibco
Boiling Point 640°C
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.