Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sparfloxacin, TCI America™
SDP

Catalog No. S089625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S089625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S089625G Supplier TCI America Supplier No. S089625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110871-86-8
Molecular Formula C19H22F2N4O3
Molecular Weight (g/mol) 392.41
MDL Number MFCD00869619
InChI Key DZZWHBIBMUVIIW-DTORHVGOSA-N
Synonym sparfloxacin, zagam, esparfloxacino, sparfloxacine, sparfloxacinum, spara, sparfloxacine inn-french, sparfloxacinum inn-latin, spfx, esparfloxacino inn-spanish
PubChem CID 60464
ChEBI CHEBI:9212
IUPAC Name 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O

Specifications

Melting Point 265°C
Color Yellow
Quantity 25 g
Formula Weight 392.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sparfloxacin
RTECSNumber VB1986500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.