Learn More
Medchemexpress LLC Sparfosic acid | 51321-79-0 | 99.3% | 255.12 g/mol | C6H10NO8P | 5 MG
SDP
Catalog No. 5000262317
Encompass_Preferred
Sparfosic acid is N-phosphonacetyl-L-aspartic acid, a DNA antimetabolite and potent inhibitor of aspartate transcarbamoyl transferase (ATCase). It is used in biochemical and cell-based research to disrupt de novo pyrimidine biosynthesis and has been reported to increase the cytotoxic effect of 5-fluorouracil combined with interferon-alpha in cancer cell models.
  • Inhibits aspartate transcarbamoyl transferase (ATCase).
  • Acts as a DNA antimetabolite affecting pyrimidine biosynthesis.
  • Reported to enhance cytotoxicity of 5-fluorouracil plus interferon-alpha.
  • High purity (99.3%) suitable for biochemical studies.
  • Available in small research pack sizes (1 mg-100 mg).
  • Molecular formula C6H10NO8P; molecular weight 255.12 g/mol.
Catalog No. 50-002-62317 Supplier Medchemexpress LLC Supplier No. HY1127325MG
Add to Cart
Add to Cart
Sparfosic acid is N-phosphonacetyl-L-aspartic acid, a DNA antimetabolite and potent inhibitor of aspartate transcarbamoyl transferase (ATCase). It is used in biochemical and cell-based research to disrupt de novo pyrimidine biosynthesis and has been reported to increase the cytotoxic effect of 5-fluorouracil combined with interferon-alpha in cancer cell models.
  • Inhibits aspartate transcarbamoyl transferase (ATCase).
  • Acts as a DNA antimetabolite affecting pyrimidine biosynthesis.
  • Reported to enhance cytotoxicity of 5-fluorouracil plus interferon-alpha.
  • High purity (99.3%) suitable for biochemical studies.
  • Available in small research pack sizes (1 mg-100 mg).
  • Molecular formula C6H10NO8P; molecular weight 255.12 g/mol.