Learn More
Bellco Glass, Inc. Sparger w/Holes,3L
SDP
Catalog No. 501944963
Encompass_Preferred
316 l stainless steel sparger assembly with drilled holes for 3-liter bioreactor vessel. MADE IN THE USA. .250x.152 Welded flange,"3/8" Silicone 'o' ring
Catalog No. 50-194-4963 Supplier Bellco Glass, Inc. Supplier No. 196497000
316 l stainless steel sparger assembly with drilled holes for 3-liter bioreactor vessel. MADE IN THE USA. .250x.152 Welded flange,"3/8" Silicone 'o' ring