Learn More
Biolegend Spark BlueTM 550 anti-mouse CD
Supplier:  Biolegend 109862
Encompass_Preferred
Spark Blue™ 550 anti-mouse CD45.2 [104]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-272-2767