Learn More
Biolegend Spark NIR™ 685 anti-human CD4
Supplier:  Biolegend 344657
Encompass_Preferred
Spark NIR™ 685 anti-human CD4 [SK3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1771566