Learn More
Biolegend Spark NIR™ 685 anti-human IFN-γ
Supplier:  Biolegend 502551
Encompass_Preferred
Spark NIR™ 685 anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-5568