Learn More
Biolegend Spark NIR™ 685 anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Supplier:  Biolegend 506359
Encompass_Preferred
Spark NIR™ 685 anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-237-4641