Learn More
Biolegend Spark NIRTM 685 anti-human CD1
Catalog No. 502721981
Encompass_Preferred
Spark NIR™ 685 anti-human CD14 [S18004B]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-1981 Supplier Biolegend Supplier No. 399210
Spark NIR™ 685 anti-human CD14 [S18004B]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests