Learn More
Biolegend Spark NIRTM 685 anti-mouse CD6
Catalog No. 502722791
Encompass_Preferred
Spark NIR™ 685 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2791 Supplier Biolegend Supplier No. 104557
Spark NIR™ 685 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 μg