Learn More
Biolegend Spark NIRTM 685 anti-mouse IFN
Catalog No. 502721886
Encompass_Preferred
Spark NIR™ 685 anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg
Catalog No. 50-272-1886 Supplier Biolegend Supplier No. 505861
Spark NIR™ 685 anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg