Learn More
Biolegend Spark PLUS B550TM anti-mouse C
Catalog No. 502906329
Encompass_Preferred
Spark PLUS B550™ anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-290-6329 Supplier Biolegend Supplier No. 104763
Spark PLUS B550™ anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 μg