Learn More
Biolegend Spark PLUS B550TM anti-mouse C
Supplier:  Biolegend 102088
Encompass_Preferred
Spark PLUS B550™ anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-290-6291