Learn More
Biolegend Spark PLUS UV™ 395 anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 502723361
Encompass_Preferred
Spark PLUS UV™ 395 anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-272-3361 Supplier Biolegend Supplier No. 307664
Spark PLUS UV™ 395 anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests