Learn More
Biolegend Spark PLUS UV™ 395 anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 502723141
Encompass_Preferred
Spark PLUS UV™ 395 anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-3141 Supplier Biolegend Supplier No. 393335
Spark PLUS UV™ 395 anti-human CD57 Recombinant [QA17A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests