Learn More
Biolegend Spark PLUS V475TM anti-mouse C
Supplier:  Biolegend 104018
Encompass_Preferred
Spark PLUS V475 anti-mouse CD8a [53-6 7] Isotype Rat IgG2a Reactivity Mouse Apps FC Size 100 g
Catalog No. 50-000-64441