Learn More
Biolegend Spark PLUS V475TM anti-mouse/h
Catalog No. 5000064429
Encompass_Preferred
Spark PLUS V475 anti-mouse/human CD11b [M1/70] Isotype Rat IgG2b Reactivity Mouse Human Cynomolgus Rhesus Apps FC Size 100 g
Catalog No. 50-000-64429 Supplier Biolegend Supplier No. 101260
Spark PLUS V475 anti-mouse/human CD11b [M1/70] Isotype Rat IgG2b Reactivity Mouse Human Cynomolgus Rhesus Apps FC Size 100 g