Learn More
Biolegend Spark Red™ 718 anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Supplier:  Biolegend 339956
Encompass_Preferred
Spark Red™ 718 anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-237-7071