Learn More
Biolegend Spark RedTM 718 anti-mouse Sig
Catalog No. 503108082
Encompass_Preferred
Spark Red 718 anti-mouse Siglec-G [SH1] Isotype Mouse IgG1 Reactivity Mouse Apps FC Size 100 g
Catalog No. 50-310-8082 Supplier Biolegend Supplier No. 163314
Spark Red 718 anti-mouse Siglec-G [SH1] Isotype Mouse IgG1 Reactivity Mouse Apps FC Size 100 g