Learn More
Biolegend Spark RedTM 718 Armenian Ham
Supplier:  Biolegend 400983
Encompass_Preferred
Spark Red™ 718 Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Apps: FC, ICFC; Size: 25 ug
Catalog No. 50-253-7822