Learn More
Biolegend Spark UVTM 387 anti-human CD66
Catalog No. 502722457
Encompass_Preferred
Spark UV™ 387 anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2457 Supplier Biolegend Supplier No. 305128
Spark UV™ 387 anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests