Learn More
Biolegend Spark UVTM 387 anti-human HL
Catalog No. 502537935
Encompass_Preferred
Spark UV™ 387 anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-253-7935 Supplier Biolegend Supplier No. 311461
Spark UV™ 387 anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests