Learn More
Biolegend Spark VioletTM 423 anti-huma
Supplier:  Biolegend 366641
Encompass_Preferred
Spark Violet™ 423 anti-human CD33 [P67.6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-253-7881