Learn More
Biolegend Spark VioletTM 423 anti-human
Catalog No. 502722342
Encompass_Preferred
Spark Violet™ 423 anti-human CD14 Recombinant [QA18A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2342 Supplier Biolegend Supplier No. 324904
Spark Violet™ 423 anti-human CD14 Recombinant [QA18A04]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests