Learn More
Biolegend Spark Violet™ 423 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 502723378
Encompass_Preferred
Spark Violet™ 423 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-3378 Supplier Biolegend Supplier No. 304055
Spark Violet™ 423 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests