Learn More
Biolegend Spark YGTM 581 anti-mouse I-A/
Catalog No. 502722777
Encompass_Preferred
Spark YG™ 581 anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2777 Supplier Biolegend Supplier No. 107665
Spark YG™ 581 anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg