Learn More
Sparkfun Electronics SparkFun Third Hand Kit
Catalog No. NC1338628
Encompass_Preferred
The SparkFun Third Hand gives you the ability to hold a board between two alligator clips on two flexible arms with multiple points of articulation. The arm's wide range of articulation gives you access to any angle of the board that you are currently working on, making soldering that much easier. The SparkFun Third Hand also won't move around while you work thanks to a heavy aluminum base supporting the arms.
Catalog No. NC1338628 Supplier Sparkfun Electronics Supplier No. TOL11784
The SparkFun Third Hand gives you the ability to hold a board between two alligator clips on two flexible arms with multiple points of articulation. The arm's wide range of articulation gives you access to any angle of the board that you are currently working on, making soldering that much easier. The SparkFun Third Hand also won't move around while you work thanks to a heavy aluminum base supporting the arms.