Learn More
Moldex™ SparkPlugs™ Ecostation™ Ear Plug Refill
SDP
Catalog No. 19384372 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
SparkPlugs™ Ecostation™ Ear Plug Refill
Catalog No.
19-384-372 SparkPlugs™ Ecostation™ Ear Plug Refill
1 options
This item is not returnable. View return policy
Catalog No. 19-384-372 Supplier Moldex™ Supplier No. 6704
Add to Cart
Add to Cart
This item is not returnable. View return policy
Convenient refill container for SparkPlugs™ Ecostation™ dispenser (6703). SparkPlugs™ are tapered to fit easily in the ear canal and made of extra-soft, extra-light, multi-colored foam.
  • Extra-soft, extra-light foam with nonirritating surface
  • Tapered for easy insertion and removal
  • Seals ear canal comfortably, without pressure
  • Disposable
  • Every package contains random multi-colored SparkPlugs
  • Provide a secure fit in all sizes of ear canals
  • Hygienically packaged
  • 100% PVC-free
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
Packaging Type Refill
Pairs per Package 500 Pairs/Dispenser Refill
Plug Material Foam
Plug Color Multi-color
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No