Learn More
Selleck Chemical LLC Sparsentan
SDP
Supplier:  Selleck Chemical LLC S66652MG
Encompass_Preferred
Sparsentan (PS-433540 RE-021 DARA) is a dual endothelin type A receptor(ETA) and angiotensin II type 1 receptor antagonist
Catalog No. 50-000-01260