Learn More
AdipoGen -Spartein Surrogate
SDP
Catalog No. 502560081
Encompass_Preferred
Chemical. CAS 475301-86-1. Formula C12H22N2. MW 194.32. Synthetic. Chiral reagent.
Catalog No. 50-256-0081 Supplier AdipoGen Supplier No. CDXS0045M100
Chemical. CAS 475301-86-1. Formula C12H22N2. MW 194.32. Synthetic. Chiral reagent.