Learn More
Medchemexpress LLC HY-W008350 100mg Medchemexpress, (+)-Sparteine CAS:492-08-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW008350100MG
Encompass_Preferred
Medchemexpress, HY-W008350 100mg (+)-Sparteine CAS:492-08-0 (+)-Sparteine is a natural alkaloid acting as a ganglionic blocking agent. (+)-Sparteine competitively blocks nicotinic ACh receptor in the neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4511