Learn More
Apexbio Technology LLC Sparteine 492-08-0 200mg
SDP
Catalog No. 5000090222
Encompass_Preferred
Sparteine (CAS 492-08-0) is a naturally occurring alkaloid recognized for its activity as a ganglionic blocker It acts by competitively inhibiting neuronal nicotinic acetylcholine receptors thereby modulating synaptic transmission within the nervous system Owing to this specific mechanism sparteine is widely employed in neurobiological and pharmacological research to investigate neuronal signaling pathways and to elucidate mechanisms involved in central nervous system disorders Its use facilitates the exploration of neurotransmitter regulation and receptor function in experimental models
Catalog No. 50-000-90222 Supplier Apexbio Technology LLC Supplier No. C6976200
Sparteine (CAS 492-08-0) is a naturally occurring alkaloid recognized for its activity as a ganglionic blocker It acts by competitively inhibiting neuronal nicotinic acetylcholine receptors thereby modulating synaptic transmission within the nervous system Owing to this specific mechanism sparteine is widely employed in neurobiological and pharmacological research to investigate neuronal signaling pathways and to elucidate mechanisms involved in central nervous system disorders Its use facilitates the exploration of neurotransmitter regulation and receptor function in experimental models