Learn More
Medchemexpress LLC (+)-Sparteine sulfate pentahydrate | 98.0% | 50 MG
SDP
Catalog No. 5000411097
Encompass_Preferred
(+)-Sparteine sulfate pentahydrate is a ganglionic blocking agent that competitively blocks nicotinic ACh receptors in neurons. This compound reduces ACh-induced current and excitatory postsynaptic currents, indicating its role in modulating neuronal activity.
  • Competitively blocks nicotinic ACh receptors in neurons.
  • Reduces ACh-induced current in a voltage-independent manner.
  • Increases blocking effect at more negative membrane potentials.
  • Reduces the amplitude of excitatory postsynaptic currents (EPSC).
  • Reduces the time constant of EPSC decay.
Catalog No. 50-004-11097 Supplier Medchemexpress LLC Supplier No. HYB1304A50MG
(+)-Sparteine sulfate pentahydrate is a ganglionic blocking agent that competitively blocks nicotinic ACh receptors in neurons. This compound reduces ACh-induced current and excitatory postsynaptic currents, indicating its role in modulating neuronal activity.
  • Competitively blocks nicotinic ACh receptors in neurons.
  • Reduces ACh-induced current in a voltage-independent manner.
  • Increases blocking effect at more negative membrane potentials.
  • Reduces the amplitude of excitatory postsynaptic currents (EPSC).
  • Reduces the time constant of EPSC decay.