Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Sparteine 98.0+%, TCI America™
SDP

Catalog No. S08841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08841G Supplier TCI America Supplier No. S08841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 492-08-0
Molecular Formula C15H26N2
Molecular Weight (g/mol) 234.387
MDL Number MFCD00869353
InChI Key SLRCCWJSBJZJBV-TUVASFSCSA-N
Synonym Pachycarpine
PubChem CID 7014
SMILES C1CCN2CC3CC(C2C1)CN4C3CCCC4

Specifications

Color Yellow
Boiling Point 174°C
Quantity 1 g
Formula Weight 234.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Sparteine
RTECSNumber RT0620000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.