Learn More
Medchemexpress LLC HY-111548 10mg Medchemexpress, Spastazoline CAS:2351882-11-4 Purity:>98%
SDP
Catalog No. 501968914
Encompass_Preferred
Medchemexpress, HY-111548 10mg Spastazoline CAS:2351882-11-4 Spastazoline is a potent and selective spastin (a microtubule-severing AAA protein) inhibitor, with an IC50 of 99 nM for Human spastin. Spastazoline shows no effect on ATPase activity of a recombinant human VPS4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8914 Supplier Medchemexpress LLC Supplier No. HY11154810MG
Medchemexpress, HY-111548 10mg Spastazoline CAS:2351882-11-4 Spastazoline is a potent and selective spastin (a microtubule-severing AAA protein) inhibitor, with an IC50 of 99 nM for Human spastin. Spastazoline shows no effect on ATPase activity of a recombinant human VPS4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.