Learn More
Aviva Systems Biology Corp SPATA16 Antibody - N-terminal region (ARP58663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPATA16. Alternate names: NYD-SP12, SPGF6ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-62390