Learn More
Aviva Systems Biology Corp SPATA2 Antibody - C-terminal region (ARP59150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SPATA2. Alternate names: FLJ13167, KIAA0757, PD1, tamoApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-596