Learn More
Aviva Systems Biology Corp SPATA2L Antibody - middle region (ARP52903_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52903_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPATA2L. Alternate names: C16orf76, MGC26885, tamoApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-60865