Learn More
Aviva Systems Biology Corp SPATA5 Antibody - middle region (ARP52815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPATA5. Alternate names: AFG2, SPAFApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60840