Learn More
Aviva Systems Biology Corp SPATA6 Antibody - middle region (ARP53736_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53736_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPATA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPATA6. Alternate names: FLJ10007, SRF-1, SRF1, HASHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53728