Learn More
Cayman Chemical Spatrgn 10mg
SDP
Catalog No. 502950628
Encompass_Preferred
A neuroprotective agent; inhibits neuronal voltage-gated sodium and calcium channels in a voltage-dependent manner; reduces the neuronal firing frequency and decreases the amplitude of EPSPs in electrically stimulated rat corticostriatal slices (EC50s = 4.5 and 2 µM, respectively); inhibits veratrine-induced glutamate and aspartate release in rat cerebral cortex slices (IC50 = ~5 µM for both); reduces infarct volume in a rat model of MCAO-induced focal ischemia at 30 mg/kg
Catalog No. 50-295-0628 Supplier Cayman Chemical Supplier No. 3296310 MG
A neuroprotective agent; inhibits neuronal voltage-gated sodium and calcium channels in a voltage-dependent manner; reduces the neuronal firing frequency and decreases the amplitude of EPSPs in electrically stimulated rat corticostriatal slices (EC50s = 4.5 and 2 µM, respectively); inhibits veratrine-induced glutamate and aspartate release in rat cerebral cortex slices (IC50 = ~5 µM for both); reduces infarct volume in a rat model of MCAO-induced focal ischemia at 30 mg/kg