Learn More
Cole-Parmer Rubbermaid 1938 NSF-Approved High-Heat Spatulas, PP/Rubber, 16-1/2"L
Catalog No. NC9415155 Shop All Cole-Parmer Products
Encompass_Preferred
Scooping, spooning, and spreading have never been easier with these spoon-shaped spatulas. The sturdy polypropylene (PP) handle is permanently bonded to a flexible rubber blade. Spatulas withstand temperatures to 212°F (100°C). NSF-approved.
Catalog No. NC9415155 Supplier Cole-Parmer Supplier No. 0627835
Scooping, spooning, and spreading have never been easier with these spoon-shaped spatulas. The sturdy polypropylene (PP) handle is permanently bonded to a flexible rubber blade. Spatulas withstand temperatures to 212°F (100°C). NSF-approved.