Learn More
Electron Microscopy Sciences Stainless Steel Spatula with Short Flat Squared and Long Flat Round Tip
SDP
Catalog No. 501930654
Encompass_Preferred
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Short and flat with squared tip and long flat with rounded tip.
TipsOAL: 165 mm
Catalog No. 50-193-0654 Supplier Electron Microscopy Sciences Supplier No. 7832614
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Short and flat with squared tip and long flat with rounded tip.
TipsOAL: 165 mm