Learn More
Chemglass Life Sciences Spatula, Micro, Minum-Ware
SDP
Catalog No. 501532676
Encompass_Preferred
Spatula, Micro, Minum-Ware. Nickel-stainless spatula with PTFE coated ends. Length is 184mm with one end tapered to 0.8mm O.D. while the opposite end is 4mm O.D. and rounded.
Catalog No. 50-153-2676 Supplier Chemglass Life Sciences Supplier No. MW13101
Spatula, Micro, Minum-Ware. Nickel-stainless spatula with PTFE coated ends. Length is 184mm with one end tapered to 0.8mm O.D. while the opposite end is 4mm O.D. and rounded.