Learn More
Andwin Scientific SPATULA MICRO SS HAYMAN 6 1/2
SDP
Catalog No. NC0619138
Encompass_Preferred
The 6.5 in. Hayman style spatula is ideal for transferring or mixing small sample amounts. It features stainless steel construction, round spatula on one end and a spoon on the other.
Catalog No. NC0619138 Supplier Andwin Scientific Supplier No. 82027522
The 6.5 in. Hayman style spatula is ideal for transferring or mixing small sample amounts. It features stainless steel construction, round spatula on one end and a spoon on the other.