Learn More
Electron Microscopy Sciences Stainless Steel Spatula with Short Pointed and Long Curved Rounded Tip
SDP
Catalog No. 501930652
Encompass_Preferred
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Pointed tip arrow-shaped and flat long curved with a rounded tip.
TipsOAL: 180 mm
Catalog No. 50-193-0652 Supplier Electron Microscopy Sciences Supplier No. 7832612
Spatulas are general purpose tools for scraping, getting into small areas, working with adhesives, and moving moving small electronic parts. Made of rust-free stainless steel AISI 410 with serrated handles for a better grip. Pointed tip arrow-shaped and flat long curved with a rounded tip.
TipsOAL: 180 mm