Learn More
Chemglass Life Sciences Spatula, Stainless Steel #304, 12" OAL, Non-Magnetic, Heavy Duty
SDP
Catalog No. CG198615
Encompass_Preferred
Spatula will pass through a 24/40 outer joint. Heavy duty, extra long spatula made of non-magnetic stainless steel. One end is rounded while the other is flat. Both ends are 3/4in (19mm) O.D. x 3in (75mm) long.
Catalog No. CG198615 Supplier Chemglass Life Sciences Supplier No. CG198615
Spatula will pass through a 24/40 outer joint. Heavy duty, extra long spatula made of non-magnetic stainless steel. One end is rounded while the other is flat. Both ends are 3/4in (19mm) O.D. x 3in (75mm) long.