Learn More
Chemglass Life Sciences Spatula, Stainless Steel #304, 6.5" OAL, Non-Magnetic, Spoon Style
SDP
Catalog No. 50873389
Encompass_Preferred
Spatula will pass through a 14/10 or 14/20 outer joint. Spoon type spatula is made of non-magnetic stainless steel with an overall length of 6.5in (165mm). The oval spoon end is 3/4in (19mm) long x 3/16in (4.8mm) wide. Equipped with a cushion grip vinyl handle for easy holding.
Catalog No. 50-873-389 Supplier Chemglass Life Sciences Supplier No. CG198312
Spatula will pass through a 14/10 or 14/20 outer joint. Spoon type spatula is made of non-magnetic stainless steel with an overall length of 6.5in (165mm). The oval spoon end is 3/4in (19mm) long x 3/16in (4.8mm) wide. Equipped with a cushion grip vinyl handle for easy holding.