Learn More
Chemglass Life Sciences Spatula, Stainless Steel #304, 8" OAL, Non-Magnetic, Rounded Ends
SDP
Catalog No. 50873388
Encompass_Preferred
Spatula will pass through a 14/10 or 14/20 outer joint. Made of non-magnetic stainless steel having a mirror finish. Both ends are rounded and are approximately 5/16" (8mm) wide x 2" (51mm) in length.
Catalog No. 50-873-388 Supplier Chemglass Life Sciences Supplier No. CG198110
Spatula will pass through a 14/10 or 14/20 outer joint. Made of non-magnetic stainless steel having a mirror finish. Both ends are rounded and are approximately 5/16" (8mm) wide x 2" (51mm) in length.